Anti-AHSG Rabbit Monoclonal Antibody [clone:012]

Supplier: Sino Biological
50093-R012-50 50093-R012-1 50093-R012-100
103594-044EA 150.18 USD
103594-044 103656-878 103588-886
Anti-AHSG Rabbit Monoclonal Antibody [clone:012]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin-A (rM Fetuin-A; Catalog#50093-M08H; NP_038493.1; Met1-Ile345).

Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 012
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR