Anti-FCGR1 Rabbit Monoclonal Antibody [clone:008]

Supplier: Sino Biological
50086-R008-20 50086-R008-100 50086-R008-50
76777-304EA 83.6 USD
76777-304 103593-996 103656-582
Anti-FCGR1 Rabbit Monoclonal Antibody [clone:008]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD64 / FCGR1 (rM CD64 / FCGR1; Catalog#50086-M08H; NP_034316.1; Met 1-Pro 297).

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 008
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR