Anti-ASGPR1 Rabbit Polyclonal Antibody

50083-RP01-400 50083-RP01-100 50083-RP01-200
103656-564EA 214.31 USD
103656-564 103656-562 103656-560
Anti-ASGPR1 Rabbit Polyclonal Antibody
Anti-Asialoglycoprotein Recept

Type: Primary
Antigen: ASGPR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR