Anti-BMPR1A Rabbit Monoclonal Antibody [clone:002]

Supplier: Sino Biological
50078-R002-100 50078-R002-50
103593-972EA 150.18 USD
103593-972 103656-542
Anti-BMPR1A Rabbit Monoclonal Antibody [clone:002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ALK-3 / BMPRIA (rM ALK-3 / BMPRIA; Catalog#50078-M08H; NP_033888.2; Met 1-Arg 152).

Type: Primary
Antigen: BMPR1A
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR