Anti-CSF1R Rabbit Polyclonal Antibody

Supplier: Sino Biological
50059-RP02-200 50059-RP02-100 50059-RP02-50
103593-852EA 142.96 USD
103593-852 103656-430 103656-428
Anti-CSF1R Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CSF1R / MCSF Receptor / CD115 (rM CSF1R / MCSF Receptor / CD115; Catalog#50059-M08H; NP_001032948.2; Met 1-Ser 511). CSF1R / MCSF Receptor / CD115 specific IgG was purified by Mouse CSF1R / MCSF Receptor / CD115 affinity chromatography.

Type: Primary
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR