Anti-SFRP4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50053-RP02-200 50053-RP02-100 50053-RP02-50
103656-404EA 270.87 USD
103656-404 103593-844 103656-406
Anti-SFRP4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse sFRP4 (rM sFRP4; Catalog#50053-M08H; NP_057896.1; Met 1-Ser 351). sFRP4 specific IgG was purified by mouse sFRP4 affinity chromatography.

Type: Primary
Antigen: SFRP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR