Anti-CCL17 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50046-RP01-200 50046-RP01-400 50046-RP01-100
103656-358EA 351.49 USD
103656-358 103656-362 103656-360
Anti-CCL17 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CCL17 / TARC (rM CCL17 / TARC; Catalog#50046-MNAB; AAD56601.1; Met1-Pro93). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR