Anti-SELL Rabbit Monoclonal Antibody [clone:414] PE (Phycoerythrin)

Supplier: Sino Biological
50045-R414-P-100 50045-R414-P-25
103631-922EA 151.5 USD
103631-922 103629-910
Anti-SELL Rabbit Monoclonal Antibody [clone:414] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SELL / CD62L / L-selectin (rM SELL / CD62L / L-selectin; Catalog#50045-M08H; NP_035476.1; Met 1-Asn 332) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: Clone: 414
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR