Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELL Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 414]

Supplier: Sino Biological

50045-R414-C-100 50045-R414-C-25
103631-918 103629-906
Anti-SELL Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 414]
Anti-SELL Antibody (PerCP)

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: Clone: 414
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty