Anti-FSTL3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50029-RP02-200 50029-RP02-100 50029-RP02-50
103656-268EA 270.87 USD
103656-268 103593-744 103656-270
Anti-FSTL3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FLRG (rM FLRG; Catalog#50029-M08H; NP_113557.1; Met 1-Val 256). FLRG specific IgG was purified by mouse FLRG affinity chromatography
.

Type: Primary
Antigen: FSTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR