Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF6 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 004]

Supplier: Sino Biological

50027-R004-F-25 50027-R004-F-100
103629-892 103632-104
Anti-TNFRSF6 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 004]
Anti-Fas Antibody (FITC)

Type: Primary
Antigen: TNFRSF6
Clonality: Monoclonal
Clone: Clone: 004
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty