Anti-FAS Rabbit Monoclonal Antibody [clone:004] APC (Allophycocyanin)

Supplier: Sino Biological
50027-R004-A-25 50027-R004-A-100
103629-890EA 30.3 USD
103629-890 103632-102
Anti-FAS Rabbit Monoclonal Antibody [clone:004] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD95 / APO-1 / TNFRSF6 / FAS (rM CD95 / APO-1 / TNFRSF6 / FAS; Catalog#50027-M08H; NP_032013.2; Met1-Arg169) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: TNFRSF6
Clonality: Monoclonal
Clone: Clone: 004
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR