Anti-CD14 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50023-RP02-50 50023-RP02-100 50023-RP02-200
103656-244EA 352.03 USD
103656-244 103656-242 103597-694
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD14 extracellular domain (rM CD14; Catalog#50023-M03H; NP_033971.1; Met 1-Pro 345). CD14 specific IgG was purified by mouse CD14 affinity chromatography.

Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR