Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD14 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50023-RP01-100 50023-RP01-200 50023-RP01-400
103656-236EA 106.62 USD InStock
103656-236 103656-238 103656-240
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
Anti-CD14 Antibody

Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty