Anti-CD14 Rabbit Monoclonal Antibody [clone:503] APC (Allophycocyanin)

Supplier: Sino Biological
50023-R503-A-25 50023-R503-A-100
103632-094EA 282.8 USD
103632-094 103629-882
Anti-CD14 Rabbit Monoclonal Antibody [clone:503] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD14 (rM CD14; Catalog#50023-M08H; NP_033971.1; Met1-Pro345) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: Clone: 503
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR