Anti-CD5L Rabbit Monoclonal Antibody [clone:38]

Supplier: Sino Biological
50020-R038-50 50020-R038-100
103656-212EA 271.4 USD
103656-212 103593-782
Anti-CD5L Rabbit Monoclonal Antibody [clone:38]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5L / Aim (rM CD5L; Catalog#50020-M08H; Met 1-Val 352; NP_033820.2)
.

Type: Primary
Antigen: CD5L
Clonality: Monoclonal
Clone: 38
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR