Anti-CXADR Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50019-R001-50 50019-R001-100
103656-200EA 271.4 USD
103656-200 103593-778
Anti-CXADR Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CXADR (rM CXADR; Catalog#50019-M08H; NP_001020363.1; Met1-Gly237).

Type: Primary
Antigen: CAR
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR