Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CA14 Rabbit Monoclonal Antibody [clone: 033]

Supplier: Sino Biological

50013-R033-50 50013-R033-100
103593-756 103656-144
Anti-CA14 Rabbit Monoclonal Antibody [clone: 033]
Anti-Carbonic Anhydrase XIV An

Type: Primary
Antigen: CA14
Clonality: Monoclonal
Clone: Clone: 033
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty