Anti-BCL2L1 Rabbit Monoclonal Antibody [clone:042]

Supplier: Sino Biological
50012-R042-50 50012-R042-100
103593-752EA 150.18 USD
103593-752 103656-132
Anti-BCL2L1 Rabbit Monoclonal Antibody [clone:042]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BCL2L1 / Bcl-XL (rM BCL2L1 / Bcl-XL; Catalog#50012-M08E; Q64373-1; Met1-Arg212).

Type: Primary
Antigen: Bcl-XL
Clonality: Monoclonal
Clone: 042
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR