Anti-ACY1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50008-RP01-100 50008-RP01-400 50008-RP01-200
103656-116EA 562.97 USD
103656-116 103656-114 103656-112
Anti-ACY1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Aminoacylase-1 / ACY1 (rM Aminoacylase-1 / ACY1; Catalog#50008-M08H; Q99JW2; Met1-Ser408). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR