Anti-ASAH2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50006-RP02-200 50006-RP02-100 50006-RP02-50
103656-108EA 270.87 USD
103656-108 103656-110 103612-502
Anti-ASAH2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ASAH2 (rM ASAH2; Catalog#50006-M07H; Thr 34-Thr 756; NP_061300.1). ASAH2 specific IgG was purified by mouse ASAH2 affinity chromatography.

Type: Primary
Antigen: ASAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR