Anti-ALCAM Rabbit Monoclonal Antibody [clone:311] APC (Allophycocyanin)

Supplier: Sino Biological
50005-R311-A-25 50005-R311-A-100
103629-872EA 111.1 USD
103629-872 103631-908
Anti-ALCAM Rabbit Monoclonal Antibody [clone:311] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD166 / ALCAM (rM CD166 / ALCAM; Catalog#50005-M08H; NP_033785.1; Met1-Lys527) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: Clone: 311
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR