Anti-ALCAM Rabbit Monoclonal Antibody [clone: 302]

50005-R302-50 50005-R302-100
103612-334EA 128.15 USD
103612-334 103656-086
Anti-ALCAM Rabbit Monoclonal Antibody [clone: 302]
Anti-CD166 / ALCAM Antibody

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: Clone: 302
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR