Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZIKV-NS1 Mouse Monoclonal Antibody [clone: 03]

Supplier: Sino Biological

40544-MM03-1 40544-MM03-50 40544-MM03-100
103588-834 103612-320 103656-070
Anti-ZIKV-NS1 Mouse Monoclonal Antibody [clone: 03]
Zika virus (ZIKV) (strain Zika

Type: Primary
Antigen: ZIKV-NS1
Clonality: Monoclonal
Clone: Clone: 03
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Zika virus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty