Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VP24 Protein Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

40456-MM01-100 40456-MM01-50
103656-058 103612-312
Anti-VP24 Protein Mouse Monoclonal Antibody [clone: 01]
Ebola virus EBOV (subtype Bund

Type: Primary
Antigen: VP24 Protein
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Ebola virus EBOV (subtype Bundibugyo;strain Uganda 2007)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty