Anti-EBOV-NP Rabbit Polyclonal Antibody

Supplier: Sino Biological
40444-T62-50 40444-T62-100
103612-296EA 312.91 USD
103612-296 103588-820
Anti-EBOV-NP Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP ( Catalog#40444-V07E1; YP_138520.1; Gly630-Asp738). Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP specific IgG was purified by Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP affinity chromatography.

Type: Primary
Antigen: Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ebola virus EBOV (subtype Zaire;strain H.sapiens-wt / GIN / 2014 / Kissidougou-C15)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR