Anti-EBOV-NP Mouse Monoclonal Antibody [clone:07]

Supplier: Sino Biological
40443-MM07-100 40443-MM07-50
103612-284EA 188.32 USD
103612-284 103656-022
Anti-EBOV-NP Mouse Monoclonal Antibody [clone:07]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP ( Catalog#40443-V07E1; AHX24646.1; His630-Gln739). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Nucleoprotein
Clonality: Monoclonal
Clone: 07
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Ebola virus EBOV (subtype Zaire;strain H.sapiens-wt / GIN / 2014 / Kissidougou-C15)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR