Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Glycoprotein Rabbit Polyclonal Antibody

Supplier: Sino Biological

40442-T24-100 40442-T24-50
103612-280 103588-810
Anti-Glycoprotein Rabbit Polyclonal Antibody
Ebola virus EBOV (subtype Zair

Type: Primary
Antigen: Glycoprotein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ebola virus EBOV (subtype Zaire;strain H.sapiens-wt / GIN / 2014 / Kissidougou-C15)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty