Anti-RVFV-L Rabbit Polyclonal Antibody

Supplier: Sino Biological
40423-RP01-200 40423-RP01-100
103656-016EA 400.2 USD
103656-016 103656-014
Anti-RVFV-L Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rift Valley fever virus (TAN/Dod-002/07) RVFV-L ( Catalog#40423-V08E; AEB20483.1; Met1-Phe250). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: RVFV-L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rift Valley fever virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR