Anti-HA Human/Mouse chimeric Monoclonal Antibody [clone:mh001]

Supplier: Sino Biological
40359-MH001-500 40359-MH001-200
103629-870EA 552.92 USD
103629-870 103629-868
Anti-HA Human/Mouse chimeric Monoclonal Antibody [clone:mh001]
Antibodies
It is a chimeric monoclonal antibody combining the constant domains of the Human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin / HA. The antibody was produced using recombinant antibody technology.

Type: Primary
Antigen: HA1
Clonality: Monoclonal
Clone: mh001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: mouse (varialbe region) / human (kappa / IgG1 constant) chimeric antibody
Reactivity: Influenza A H10N8
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR