Anti-HA Mouse Monoclonal Antibody [clone:03]

Supplier: Sino Biological
40324-MM03-100 40324-MM03-50
103612-472EA 188.32 USD
103612-472 103655-992
Anti-HA Mouse Monoclonal Antibody [clone:03]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H18N11 (A/flat-faced bat/Peru/033/2010) Hemagglutinin / HA1 ( Catalog#40324-V08H1; AGX84934.1; Met1-Arg339). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: HA1
Clonality: Monoclonal
Clone: 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Influenza A H18N11
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR