Anti-pfHPRT Rabbit Polyclonal Antibody

Supplier: Sino Biological
40301-RP01-100 40301-RP01-400 40301-RP01-200
103655-968EA 300.34 USD
103655-968 103655-970 103655-972
Anti-pfHPRT Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant P. falciparum pfHPRT (Catalog#40301-N07E; XP_001349989.1; Met1-Leu231). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: pfHPRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: P. falciparum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR