Anti-DENV-NS1 Mouse Monoclonal Antibody [clone:2]

Supplier: Sino Biological
40263-MM02-100 40263-MM02-50
103612-124EA 188.32 USD
103612-124 103655-956
Anti-DENV-NS1 Mouse Monoclonal Antibody [clone:2]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C) NS1 ( Catalog#40263-V07H; AAC59275.1; Asp1-Ala352). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: DENV
Clonality: Monoclonal
Clone: 2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR