Anti-gp120 Rabbit Polyclonal Antibody

Supplier: Sino Biological
40259-T60-100 40259-T60-50
103588-772EA 170 USD
103588-772 103612-118
Anti-gp120 Rabbit Polyclonal Antibody
Antibodies
HIV-1(group P, strain RBF168)

Type: Primary
Antigen: gp120
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: HIV1
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR