Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIV-1 p24 Protein Rabbit Polyclonal Antibody

Supplier: Sino Biological

40256-RP01-200 40256-RP01-100
103655-952 103655-950
Anti-HIV-1 p24 Protein Rabbit Polyclonal Antibody
HIV-1 p24 Protein (group O, st

Type: Primary
Antigen: HIV-1 p24 Protein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: HIV1

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty