Anti-Gag-p24 Rabbit Polyclonal Antibody

Supplier: Sino Biological
40252-RP01-100 40252-RP01-200
103655-942EA 300.55 USD
103655-942 103655-944
Anti-Gag-p24 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant HIV-1 p24 Protein (group M, subtype D, strain NDK) (Catalog#40252-V08E; AAA44868.1; Pro133-Leu363). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: HIV-1 p24 Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: HIV1
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR