Anti-HIV-1 p24 Protein Rabbit Polyclonal Antibody

Supplier: Sino Biological
40243-T24-50 40243-T24-100
103588-760EA 170 USD
103588-760 103612-106
Anti-HIV-1 p24 Protein Rabbit Polyclonal Antibody
Antibodies
HIV-1 p24 (group M, subtype B,

Type: Primary
Antigen: HIV-1 p24 Protein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: HIV1
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR