Anti-NP Rabbit Polyclonal Antibody

Supplier: Sino Biological
40207-T60-100 40207-T60-50
103612-096EA 290 USD
103612-096 103588-752
Anti-NP Rabbit Polyclonal Antibody
Antibodies
Influenza A H3N2 (A/Aichi/2/19

Type: Primary
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Influenza A H3N2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR