Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

40158-T54-100 40158-T54-50
103612-462 103588-744
Anti-HA1 Rabbit Polyclonal Antibody
Influenza H5N1 (A/chicken/Viet

Type: Primary
Antigen: HA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty