Anti-Spike S1 Rabbit Polyclonal Antibody

40150-RP01-100 40150-RP01-200
103656-856EA 247.69 USD
103656-856 103656-858
Anti-Spike S1 Rabbit Polyclonal Antibody
Human SARS Coronavirus Spike S

Type: Primary
Antigen: Spike S1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: SARS Coronavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR