Anti-NP Rabbit Polyclonal Antibody

40143-T60-50 40143-T60-100
103612-086EA 312.31 USD
103612-086 103588-092
Anti-NP Rabbit Polyclonal Antibody
Human SARS Coronavirus Nucleop

Type: Primary
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: SARS Coronavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR