Anti-HA Mouse Monoclonal Antibody [clone:06]

Supplier: Sino Biological
40103-MM06-50 40103-MM06-100
103612-076EA 188.32 USD
103612-076 103655-888
Anti-HA Mouse Monoclonal Antibody [clone:06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant H7N9 HA (Catalog#40103-V08B; EPI439507; Met1-Val524). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: H7N9
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Influenza A H7N9
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR