Anti-EBOV-G Rabbit Polyclonal Antibody

Supplier: Sino Biological
40094-RP01-200 40094-RP01-100
103655-884EA 300.55 USD
103655-884 103655-886
Anti-EBOV-G Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Sudan ebolavirus EBOV-G / Glycoprotein (Catalog#40094-V08B1; YP_138523.1; Met1-Asn637). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EBOV-G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Sudan ebolavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR