Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Spike Protein Rabbit Monoclonal Antibody [clone: 723]

Supplier: Sino Biological

40069-R723-200 40069-R723-500
103655-868EA 215.38 USD InStock
103655-868 103655-870
Anti-Spike Protein Rabbit Monoclonal Antibody [clone: 723]
Antibodies
MERS-CoV (NCoV / Novel coronav

Type: Primary
Antigen: Spike Protein
Clonality: Monoclonal
Clone: Clone: 723
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: MERS-CoV

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty