Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-T4 DNA ligase Rabbit Polyclonal Antibody

Supplier: Sino Biological

40066-RP01-200 40066-RP01-400 40066-RP01-100
103655-840 103655-844 103655-842
Anti-T4 DNA ligase Rabbit Polyclonal Antibody
Anti-T4 DNA ligase Antibody

Type: Primary
Antigen: T4 DNA ligase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: E. coli

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty