Anti-HA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
40058-T38-100 40058-T38-50
103588-672EA 170 USD
103588-672 103612-442
Anti-HA1 Rabbit Polyclonal Antibody
Antibodies
Influenza H3N2 (A/reassortant/

Type: Primary
Antigen: HA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Influenza H3N2 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934))
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR