Anti-Spike Mouse Monoclonal Antibody [clone:07]

Supplier: Sino Biological
40021-MM07-100 40021-MM07-50
103612-052EA 136.35 USD
103612-052 103655-818
Anti-Spike Mouse Monoclonal Antibody [clone:07]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 (Catalog#40021-V08H; YP_173238.1; Met 1-Arg 760). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Glycoprotein
Clonality: Monoclonal
Clone: 07
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR