Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STX2B Mouse Monoclonal Antibody [clone: 08]

Supplier: Sino Biological

40019-MM08-200 40019-MM08-1 40019-MM08-50 40019-MM08-100
103655-802 103588-660 103612-046 103655-800
Anti-STX2B Mouse Monoclonal Antibody [clone: 08]
Anti-Shiga toxin II subunit B

Type: Primary
Antigen: STX2B
Clonality: Monoclonal
Clone: Clone: 08
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Enterohemorrhagic E. coli (EHEC) stx2B;Shigella

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty