Anti-Neuraminidase Rabbit Polyclonal Antibody

Supplier: Sino Biological
40018-T56-50 40018-T56-100
103612-044EA 290 USD
103612-044 103588-658
Anti-Neuraminidase Rabbit Polyclonal Antibody
Antibodies
Influenza A H5N1 (A/Hubei/1/20

Type: Primary
Antigen: Neuraminidase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Influenza A H5N1 (A/Hubei/1/2010) Neuraminidase / NA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR