Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

40016-T38-50 40016-T38-100
103588-652 103612-038
Anti-HA1 Rabbit Polyclonal Antibody
Influenza B (B/Brisbane/60/200

Type: Primary
Antigen: HA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Influenza B (B/Brisbane/60/2008) Hemagglutinin / HA1

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty